PL APO MACRO OBJ 8X/0.22 1 * 1 items
Catalog # LEIC11581097
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
VWR®, Combs for PerfectBlue Gel System Mini L
Supplier: VWR Collection
Density Meters with Peltier Temperature Control, DSG
Supplier: BELLINGHAM STANLEY
UV/Visible spectrophotometers, DR6000
Supplier: Hach
BOD bottle, 500 mL, brown 1 * 1 items
Supplier: Hach
Accessories for measuring magnifiers
Supplier: ESCHENBACH OPTIK
REFRACTOMETER GLYCOL 1 * 1 items
Supplier: Lovibond
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Pump tubing for AKTA flux
Supplier: Cytiva
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Accessories for conductivity meters, Portavo
Supplier: KNICK
Accessories for empty columns XK series
Supplier: Cytiva
Titrator, Titronic® 300
Supplier: SI Analytics
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
Cooled incubators, FOC connect
Supplier: VELP SCIENTIFIC
Storage phosphor screens and cassettes
Supplier: Cytiva