10 L CONTAINER OF NLL NEUTRALIZING ACID 1 * 10 L
Catalog # LANC41010014
Supplier: GETINGE LIFE SCIENCES
10 L CONTAINER OF NLL NEUTRALIZING ACID 1 * 10 L
Catalog # LANC41010014
Supplier: GETINGE LIFE SCIENCES
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dishwashing liquid, green care®, MANUDISH original, Tana Professional
Supplier: WERNER MERTZ
VWR® Bulb Pipettes, Two Marks, Class AS
Supplier: VWR Collection
Bottles, Wide Neck, with Screw Cap
Supplier: GLASWARENFABRIK KARL HECHT
VWR®, Disposable Weighing and Drying Dishes or Pans
Supplier: VWR Collection
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Finntip™ Filtered Pipette Tips
Supplier: Thermo Fisher Scientific
Membrane filters, nylon, Whatman™
Supplier: Whatman products (Cytiva)
Disodium hydrogen phosphate dihydrate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Jerricans, narrow neck, HDPE, transparent or blue
Supplier: BURKLE
LLL liquid detergent
Supplier: GETINGE LIFE SCIENCES
Bottles, wide neck, with screw cap, series 303
Supplier: Kautex
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Arsenic limit test apparatus
Supplier: DIXON SCIENTIFIC
2,4-Di-tert-butylphenol 97%
Supplier: Thermo Fisher Scientific
Nitrile gloves, TouchNTuff® 92-665 / 92-670
Supplier: Ansell
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
VWR®, Roller Bottles
Supplier: VWR Collection