Masterflex® I/P® Variable-Speed Air-Powered Drive with Easy-Load® ATEX Pump Head, 100 to 650 rpm
Catalog # MFLX77980-10
Supplier: Avantor Fluid Handling
CE Compliant
Specifications
- Maximum Flow Rate:8000 ml/min
- Accuracy:±10%
- Control type:Variable-Speed Air-Powered
- Min. flow rate:1200 ml/min
- No. of channels:1
- Pump series:I/P
- Description:I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm.
- Height:22,86 cm
- Length:33,35 cm
- Seq:02
- Environmentally Preferable:
- Model:77980-10
- Duty cycle:Continuous
- Drive IP rating:IP 34
- Min. RPM:100 rpm
- Open-head sensor:No
- Tubing sizes accepted:I/P® 26, 73, 82
- Washdown capable:No
- Tubing:06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
- Drive:07589-30 (I/P® Variable-Speed Air-Powered Drive)
- No. of rollers:3
- Warranty:2 years
- Pk:1
- Pump head:77602-80 (I/P® Easy-Load® ATEX Head)
- Width:21,59 cm
- Housing material:Painted steel
- Max. RPM:650 rpm
Specifications
About this item
Pump safely in hazardous
environments.
- Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
- Easily adjust motor speed by using included regulator
- Effortlessly connect to your existing air supply or compressor
- The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
- ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6
Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.
- Maximum Flow Rate:8000 ml/min
- Accuracy:±10%
- Control type:Variable-Speed Air-Powered
- Min. flow rate:1200 ml/min
- No. of channels:1
- Pump series:I/P
- Description:I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm.
- Height:22,86 cm
- Length:33,35 cm
- Seq:02
- Environmentally Preferable:
- Model:77980-10
- Duty cycle:Continuous
- Drive IP rating:IP 34
- Min. RPM:100 rpm
- Open-head sensor:No
- Tubing sizes accepted:I/P® 26, 73, 82
- Washdown capable:No
- Tubing:06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
- Drive:07589-30 (I/P® Variable-Speed Air-Powered Drive)
- No. of rollers:3
- Warranty:2 years
- Pk:1
- Pump head:77602-80 (I/P® Easy-Load® ATEX Head)
- Width:21,59 cm
- Housing material:Painted steel
- Max. RPM:650 rpm
Frequently Bought Together