Masterflex® USB to DB9 (M) RS-232 Serial Adapter Cable
MFLX22050-58
Optimize performance of your Masterflex® pump.
- Connect your Masterflex® drive via DB9 (M) RS-232 connection to USB
Use this cable to connect the Masterflex Computer Compatible drives 07550- and 07551- series to a PC with USB port. Will also connect other peripheral devices equipped with RS-232 serial interface including balances, printers, scanners, and PDA's. Includes 2-ft cable and driver software. Supports data transfer speeds up to 12 Mbps; compatible with Windows® 2000 / XP / Vista / 7.
- Description:USB to DB9 (M) RS-232 serial adapter cable
- Environmentally Preferable:
- Model:22050-58
- Warranty:1 year
- Pk:1
Frequently Bought Together