Order Entry
United Kingdom
ContactUsLinkComponent
1631C3 Connecting Cable, Single-Core Coax, High Insulation for Sensor, KIAG 10-32 neg. KIAG 10-32 pos. - BNC pos. Length: l = 3 m D eg. of protection: IP65 - IP40 Cable PFA, High-Temperature, ø2 mm 1 * 1 items
  MSPP18000527
1631C3 Connecting Cable, Single-Core Coax, High Insulation for Sensor, KIAG 10-32 neg. KIAG 10-32 pos. - BNC pos. Length: l = 3 m D eg. of protection: IP65 - IP40 Cable PFA, High-Temperature, ø2 mm 1 * 1 items
  MSPP18000527
Frequently Bought Together