Dichloro[rel-[N(S)]-N-[2-(diphenylphosphinyl-κ O)ethyl]-2-[(R)-benzylthio-κ S]ethanamine-κ N](triphenylphosphine) ruthen ium 1 * 50 mg
Catalog # STRE44-3245-50MG
Supplier: Strem Chemicals, Inc.
Dichloro[rel-[N(S)]-N-[2-(diphenylphosphinyl-κ O)ethyl]-2-[(R)-benzylthio-κ S]ethanamine-κ N](triphenylphosphine) ruthen ium 1 * 50 mg
Catalog # STRE44-3245-50MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals