Order Entry
United Kingdom
ContactUsLinkComponent
Minitron double stack shakers without cooling. 50mm diameter throw 120mm base Temp ambient +5 to 65 Speed range 20-400 rpm Power supply 230V CO2 control 0-20% Direct steam humidification + 5L reservoir with predrilled tray for clamps. 1 * 1 items
Catalog # INFOAN121847
Supplier:  INFORS
Minitron double stack shakers without cooling. 50mm diameter throw 120mm base Temp ambient +5 to 65 Speed range 20-400 rpm Power supply 230V CO2 control 0-20% Direct steam humidification + 5L reservoir with predrilled tray for clamps. 1 * 1 items
Catalog # INFOAN121847
Supplier:  INFORS
Frequently Bought Together