Order Entry
United Kingdom
ContactUsLinkComponent
LCGC Certified Amber Glass 15 x 45 mm Screw Neck Vial, with Cap and Preslit PTFE/Silicone Septum 4 mL Volume 1 * 100 Vial
Catalog # WATE186001134C
Supplier:  WATERS
LCGC Certified Amber Glass 15 x 45 mm Screw Neck Vial, with Cap and Preslit PTFE/Silicone Septum 4 mL Volume 1 * 100 Vial
Catalog # WATE186001134C
Supplier:  WATERS
Frequently Bought Together