Additional specification - µS/cm 5.0 Max, pH 5.0 to 8.0, Sodium 50 µg/L Max, Chloride 50 µg/L Max 1 * 5 L
DIUKWA11600-275COA
Additional specification - µS/cm 5.0 Max, pH 5.0 to 8.0, Sodium 50 µg/L Max, Chloride 50 µg/L Max 1 * 5 L
DIUKWA11600-275COA
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection