JERRYCAN UN 10 LITRE NO CAP 1 * 100 items
Catalog # VWRL080513
Supplier: LAB3 OWN PRODUCTION
Frequently Bought Together
Accessories for pipette controller, BRAND accu-jet® S
Supplier: Brand
Brushes for tubes and barrels
Supplier: Socorex Isba
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Screw Neck Vials ND15
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Magnetic Stirring Bars, Cross-Shaped
Supplier: VWR Collection
Double spatulas, micro
Supplier: RSG
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Spare Parts for VWR® Hitachi HPLC Instruments
Supplier: VWR Collection
CAP DIN 51 BLACK T/EVIDENT 1 * 100 items
Supplier: LAB3 OWN PRODUCTION
Buffer solutions in sachets
Supplier: Hanna
GLOVE ALPHATEC 0.35/L320MM YELLOW 9 1 * 12 PAIR
Supplier: Ansell
pH electrodes, SenTix® PLUS
Supplier: WTW
Tape dispensers
Supplier: MICRONOVA