Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR® Septa ND8

VWR® Septa ND8

Red rubber/beige PTFE, 45° shore A, 1,0 mm
Catalog # 548-3344A
Supplier:   VWR Collection
VWR® Septa ND8
Catalog # 548-3344A
Supplier:  VWR Collection
VWR® Septa ND8
VWR® Septa ND8

Red rubber/beige PTFE, 45° shore A, 1,0 mm

Price
£157.00
Log in for availability
1 * 1.000 items
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Specifications
  • Hardness:
    45° shore A
  • Septum:
    Red rubber/beige PTFE
  • Seq:
    06
  • Thickness:
    1,0 mm