Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
VWR® Septa ND8

VWR® Septa ND8

PTFE virginal, 53° shore D, 0,25 mm
Catalog # 548-0027A
Supplier:   VWR Collection
VWR® Septa ND8
Catalog # 548-0027A
Supplier:  VWR Collection
VWR® Septa ND8
VWR® Septa ND8

PTFE virginal, 53° shore D, 0,25 mm

Price
£46.90
Log in for availability
1 * 1.000 items
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Specifications
  • Hardness:
    53° shore D
  • Septum:
    PTFE virginal
  • Seq:
    02
  • Thickness:
    0,25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.