Order Entry
United Kingdom
ContactUsLinkComponent
Measurement Capsule - Cylinder with external treads Cap 1: Sentronic NIR probe attachment for contact measuring Cap 2: Hudson Tor nado probe attachment for non-contact measuring Cap 3: Plunger with weight plate for compressing with standard weight 1 * 1 items
Catalog # MSPPMEASURECAPS
Measurement Capsule - Cylinder with external treads Cap 1: Sentronic NIR probe attachment for contact measuring Cap 2: Hudson Tor nado probe attachment for non-contact measuring Cap 3: Plunger with weight plate for compressing with standard weight 1 * 1 items
Catalog # MSPPMEASURECAPS
Frequently Bought Together