Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Bromo-4-chlorophenol 97%

2-Bromo-4-chlorophenol 97%

Bottle, 500 g
Catalog # APOSOR2160-500G
Irritant
Supplier:   Apollo Scientific
2-Bromo-4-chlorophenol 97%
Catalog # APOSOR2160-500G
Supplier:  Apollo Scientific
CAS Number:  695-96-5
2-Bromo-4-chlorophenol 97%

Bottle, 500 g

CAS Number:  695-96-5
Price
£192.00
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:695-96-5
  • Formula:C₆H₄BrClO
  • MDL Number:MFCD00002319
  • Molecular Weight:207.45 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.