2-Chlorothioxanthone 98%
Bottle, 100 g
Catalog # APOSOR471609-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
- CAS Number:86-39-5
- Formula:C₁₃H₇ClOS
- MDL Number:MFCD00005067
- Molecular Weight:246.72 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



