Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4-Methoxyquinoline 98%

4-Methoxyquinoline 98%

Bottle, 5 g
Catalog # APOSOR951081-5G
Supplier:   Apollo Scientific
4-Methoxyquinoline 98%
Catalog # APOSOR951081-5G
Supplier:  Apollo Scientific
CAS Number:  607-31-8
4-Methoxyquinoline 98%

Bottle, 5 g

CAS Number:  607-31-8
Price
£172.00
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:607-31-8
  • Formula:C₁₀H₉NO
  • MDL Number:MFCD00483884
  • Molecular Weight:159.185 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications