Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4-Methoxyquinoline 98%

4-Methoxyquinoline 98%

Bottle, 1 g
Catalog # APOSOR951081-1G
Supplier:   Apollo Scientific
4-Methoxyquinoline 98%
Catalog # APOSOR951081-1G
Supplier:  Apollo Scientific
CAS Number:  607-31-8
4-Methoxyquinoline 98%

Bottle, 1 g

CAS Number:  607-31-8
Price
£35.20
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:607-31-8
  • Formula:C₁₀H₉NO
  • MDL Number:MFCD00483884
  • Molecular Weight:159.185 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.