Order Entry
United Kingdom
ContactUsLinkComponent
Tempod® 200XB Bluetooth® Temperature Data Logger -200°C ... 70°C, External RTD sensor probe (1m), Multi-use, Bluetooth® 4.0, USB, L CD, Auto PDF report, Factory 3-point calibration, Software-free configuration 1 * 1 items
Catalog # MSPPPD200XB
Tempod® 200XB Bluetooth® Temperature Data Logger -200°C ... 70°C, External RTD sensor probe (1m), Multi-use, Bluetooth® 4.0, USB, L CD, Auto PDF report, Factory 3-point calibration, Software-free configuration 1 * 1 items
Catalog # MSPPPD200XB
Frequently Bought Together