Correx Box, 350x270x65mm Correx Box, Die cut 3mm/450gsm green Correx box FEFCO 0427. Internal dimensions @ 350 x 270 x 65mm. Low den se white foam loose in base & glued in lid at 10mm depth, Includes C13558/3/A+C13558/3/B 1 * 1 SET
Catalog # MSPPOFC13558/3
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together