Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Everolimus, Reference Standards

Everolimus, Reference Standards

Everolimus System Suitability Mixture, USP reference standard, Vial, 25 mg
Catalog # 1268987.
Supplier:   USP
Everolimus, Reference Standards
Catalog # 1268987.
Supplier:  USP
CAS Number:  
Everolimus, Reference Standards

Everolimus System Suitability Mixture, USP reference standard, Vial, 25 mg

CAS Number:  
Price
£2,680.00
Log in for availability
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Everolimus System Suitability Mixture: This is a mixture of everolimus, everolimus oxepane isomer and 9-O-hydroxyethyl sirolimus

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    USP reference standard
  • Description:
    Everolimus System Suitability Mixture
  • UN No.:
    3077
  • Cat. No.:
    1268987.
  • Description:
    Everolimus

Specifications