CELLACA MX HIGH SPEED CELL COUNTER BASE UNIT. BRIGHTFIELD IMAGING INSTRUMENT, COMPUTER CONTROLLER (WINDOWS 10), USB CABLE, POWER SUP PLY. INCLUDES A SLEEVE OF 10 COUNTING PLATES. 12 MONTH WARRANTY 1 * 1 items
Catalog # NEXCMX-BF
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together