Order Entry
United Kingdom
ContactUsLinkComponent
Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 25 µG
Catalog # ACRBSIA-H82E0-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 25 µG
Catalog # ACRBSIA-H82E0-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together