Order Entry
United Kingdom
ContactUsLinkComponent
Human SIRP alpha, Mouse IgG1 Fc Tag (HPLC-verified) (SIA-H52A8) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 1 mg
Catalog # ACRBSIA-H52A8-1MG
Supplier:  ACROBIOSYSTEMS
Human SIRP alpha, Mouse IgG1 Fc Tag (HPLC-verified) (SIA-H52A8) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession NP001035111). 1 * 1 mg
Catalog # ACRBSIA-H52A8-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together