Human SIRP alpha, His Tag (HPLC-verified) (SIA-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (A ccession NP001035111). 1 * 100 µG
Catalog # ACRBSIA-H5225100UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together