Order Entry
United Kingdom
ContactUsLinkComponent
Mouse SIRP alpha, Fc Tag (SIA-M5258) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession BAA13520 .1). 1 * 1 mg
Catalog # ACRBSIA-M5258-1MG
Supplier:  ACROBIOSYSTEMS
Mouse SIRP alpha, Fc Tag (SIA-M5258) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession BAA13520 .1). 1 * 1 mg
Catalog # ACRBSIA-M5258-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together