Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Rabbit recombinant Complement C5 (from HEK293 cells)

Rabbit recombinant Complement C5 (from HEK293 cells)

100 µG
Catalog # ACRBCO5-R52H5100UG
Marketsource
Supplier:   ACRO Biosystems
Rabbit recombinant Complement C5 (from HEK293 cells)
Catalog # ACRBCO5-R52H5100UG
Supplier:  ACRO Biosystems
Marketsource
CAS Number:  
Rabbit recombinant Complement C5 (from HEK293 cells)

100 µG

CAS Number:  
Price
£350.00
Log in for availability
1 * 100 µG
Product Details & Documents

Rabbit Complement C5 protein, His Tag.

Specifications

Specifications

  • Pk:
    100 µG
  • Protein/peptide name:
    Complement C5
  • Protein synonyms:
    CPAMD4|Complement C5|C5
  • Purity:
    >90% as determined by SDS-PAGE.
  • Endotoxin level:
    Low
  • Source:
    HEK293 cells
  • Molecular weight:
    73,8 kDa (β chain) and 111,9 kDa (α chain)
  • Shipping temperature:
    RT
  • Cat. No.:
    ACRBCO5-R52H5100UG
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Storage conditions:
    –20 °C
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Conjugation:
    Unconjugated
  • Tag sequence:
    His Tag
  • Species:
    Rabbit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.