9050105 Petri dishes, 65 x 14 mm, with triple vents, with compartments, sterilized 500pk 1 * 500 items
RATI9050105
: RATIOLAB
9050105 Petri dishes, 65 x 14 mm, with triple vents, with compartments, sterilized 500pk 1 * 500 items
RATI9050105
: RATIOLAB
Frequently Bought Together
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
VWR®, Water Sampling Bottles, Sterile
: VWR Collection
One-way pipette, ParasiTrap®
: Biosepar
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR®, Straw Pipettes
: VWR Collection
VWR®, Test Tubes without Rim
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR® Cryoboxes and Dividers
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR® Cellulose Stoppers
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF