CELLACA MX HIGH SPEED CELL COUNTER EQUIPED WITH FOM’S FOR AOPI ANALYSIS. BRIGHTFIELD IMAGING INSTRUMENT, FLUORESCENCE OPTICAL PLATFORM, GREEN AND RED CHANNELS, COMPUTER CONTROLLER (WINDOWS 10), USB CABLE, POWER SUPPLY. INCLUDES A SLEEVE OF 10 COUNTIN 1 * 1 items
Catalog # NEXCMX-AOPI
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together