Order Entry
United Kingdom
ContactUsLinkComponent
Standard profile, 0.2 ml frosted polypropylene wells, clear polycarbonate frame, cut corner A12, designed for use on standard thermal cyclers including ABI® instruments 1 * 50 items
Catalog # FTIT4TI-0772
Supplier:  4titude Ltd.
Standard profile, 0.2 ml frosted polypropylene wells, clear polycarbonate frame, cut corner A12, designed for use on standard thermal cyclers including ABI® instruments 1 * 50 items
Catalog # FTIT4TI-0772
Supplier:  4titude Ltd.
Frequently Bought Together