: FluidX 48-format, 1.9ml External Thread, Next-Gen Jacket Tri-coded Cryo Tube, 2D Code on base, 1D Linear Barcode and Human Readabl e Number on side, capped, bulk 1 * 480 items
Catalog # MSPP65-7532
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together