Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EP Reference Solution Y3, Yellow Reference Solution 1 * 120 mL
Catalog # RICCR2883300-120C
Supplier:
RICCA CHEMICAL
Certificates
Expand image
EP Reference Solution Y3, Yellow Reference Solution 1 * 120 mL
Catalog # RICCR2883300-120C
Supplier:
RICCA CHEMICAL
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Phosphate standard solution, 1000 mg/L PO₄³⁻ in water (from KH₂PO₄) Certipur®, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, ready to use, 1 litre bottles, Supelco®
Supplier: Merck