Mouse Cage for Conditioned Place Preference (CPP), including 2 context kits. Custom model for pain tests: the drawer and floor assem bly is detachable, to allow positioning the cage over a grid 37450-005 for plantar stimulation 1 * 1 items
Catalog # MSPP42553-D01
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together