Recombinant Proteinase K
Plastic bottle for solids, 100 mg
GHS symbols: GHS07; GHS08
GHS signal word: Danger
EINECS: 254-457-8
- Pack type:Plastic bottle for solids
- Pk:100 mg
- Cat. No.:A7932.0100PE
- Enzyme type:Recombinant
- Storage conditions:Freezer (2...8 °C)
- Enzyme synonyms:Recombinant Proteinase K, Recombinant Protease K, Recombinant Endopeptidase K, Recombinant Tritirachium alkaline proteinase, Recombinant Tritirachium album serine proteinase
- Enzyme name:Proteinase K
- Molecular weight:28,8 kD
- CAS No.:39450-01-6



