Order Entry
United Kingdom
ContactUsLinkComponent
Detect Human Adiponectin/ADIPOQ with <60pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cu lture supernates, serum, plasma(heparin, EDTA)and urine. 1 * 1 KIT
Catalog # BSBTFEK0595
Supplier:  Boster Bio
Detect Human Adiponectin/ADIPOQ with <60pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cu lture supernates, serum, plasma(heparin, EDTA)and urine. 1 * 1 KIT
Catalog # BSBTFEK0595
Supplier:  Boster Bio
Frequently Bought Together