Order Entry
United Kingdom
ContactUsLinkComponent
Chemsplash 2523 PolyProp Coverall Blue 2XL, Polypropylene Dust Coverall, 30 GSM non woven material, zip front closure, elasticated wrist and ankle, hood. Individually packed for hygiene purposes suitable for food processing and general non hazardous 1 * 50 items
Catalog # DIUK2523.300.008
Supplier:  DISTRIBUTOR
Chemsplash 2523 PolyProp Coverall Blue 2XL, Polypropylene Dust Coverall, 30 GSM non woven material, zip front closure, elasticated wrist and ankle, hood. Individually packed for hygiene purposes suitable for food processing and general non hazardous 1 * 50 items
Catalog # DIUK2523.300.008
Supplier:  DISTRIBUTOR
Frequently Bought Together