Order Entry
United Kingdom
ContactUsLinkComponent
Polystyrene Calibration Films for PhEur 2.2.24 Calibration of IR & FTIR Instruments 1 * 1 PAIR
Catalog # INCR0009-8158
Polystyrene Calibration Films for PhEur 2.2.24 Calibration of IR & FTIR Instruments 1 * 1 PAIR
Catalog # INCR0009-8158
Frequently Bought Together