Alum Adjuvant is insoluble white colloidal suspension of aluminum hydroxide used to potentiate immune response. Antigen adsorbs on t he aluminum hydroxide precipitate and when injected form a depot of insoluble antigen at injection site which attract 1 * 50 mL
Catalog # GENO786-1215
Supplier: G-Biosciences
Frequently Bought Together