volume 20 mL, clear glass vial (flat top), closure type, crimp top vial, O.D. × H 22.5 mm × 75.5 mm, 1 * 100 items
SIALSU860104
: Merck
volume 20 mL, clear glass vial (flat top), closure type, crimp top vial, O.D. × H 22.5 mm × 75.5 mm, 1 * 100 items
SIALSU860104
: Merck
Frequently Bought Together
VWR®, Manual Crimper and Decapper
: VWR Collection
VWR®, Short Thread Vials ND9
: VWR Collection
Exhaust filters, V3.0
: S.C.A.T.
VWR®, Snap Cap Vials
: VWR Collection
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
VWR®, Crimp Neck Vials ND11
: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
: VWR Collection
Air valves for safetycaps generation 2.0
: S.C.A.T.
VWR® Standard Micro Syringes
: VWR Collection
Caps for Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, SureSTART™
: Thermo Fisher Scientific