3,3'-BIS(3,5-DI-TERT-BUTYL-4-HYDROXYPHENYL)-N ,N'-HEXAMETHYLENEDIPROPIONAMIDE 1 * 5 g
COBBQA-2504-5G
3,3'-BIS(3,5-DI-TERT-BUTYL-4-HYDROXYPHENYL)-N ,N'-HEXAMETHYLENEDIPROPIONAMIDE 1 * 5 g
COBBQA-2504-5G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals