Order Entry
United Kingdom
ContactUsLinkComponent
Stainless steel, double platform rack 60h x 133 x 555 including handgrip with 115 17mm dia holes in a format 5 x 23. For 15 ml Falcon tubes. 1 * 1 items
  WESBDP17115HGSS
 :  WESBART
Stainless steel, double platform rack 60h x 133 x 555 including handgrip with 115 17mm dia holes in a format 5 x 23. For 15 ml Falcon tubes. 1 * 1 items
  WESBDP17115HGSS
 :  WESBART
Frequently Bought Together