2400 BOND(TM) TITRATION INSERTS 50X 1 * 1 items
Catalog # LEICOPT9719
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
Screw caps for wide neck bottles
Supplier: Kautex
Accessories for Mettler balances
Supplier: Mettler - Toledo
Labels, hazardous substance, GHS pictograms
Supplier: Brady
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Cell production roller apparatus, modular, with all position drive, Sci/ERA™
Supplier: BELLCO GLASS
Accessories for titrators
Supplier: Mettler - Toledo
BOND DEWAX SOLUTION 1 * 1 L
Supplier: LEICA MICROSYSTEMS
BOND UNIVERSAL COVERTILES 1 * 160 items
Supplier: LEICA MICROSYSTEMS
BOND TITRATION KIT 1 * 1 KIT
Supplier: LEICA MICROSYSTEMS
Accessories for measuring magnifiers
Supplier: ESCHENBACH OPTIK
BOND EPIYOPE RETRIEVAL SOLUTION 1 1 * 1 L
Supplier: LEICA MICROSYSTEMS
BOND POLYMER REFINE RED DETECTION 1 * 1 KIT
Supplier: LEICA MICROSYSTEMS
Anti-VGLUT2 Mouse Monoclonal Antibody [clone: 8G9.2]
Supplier: Abcam
BOND PRIMARY ANTIBODY DILUENT 1 * 500 mL
Supplier: LEICA MICROSYSTEMS