Order Entry
United Kingdom
ContactUsLinkComponent
Precision therm., similar ISO 655, long, 0+600:2°C, enclosed scale, cap. colourless, gallium filled, total immersion, max. 375x9mm, richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARN65094-GA
Supplier:  Amarell
Precision therm., similar ISO 655, long, 0+600:2°C, enclosed scale, cap. colourless, gallium filled, total immersion, max. 375x9mm, richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARN65094-GA
Supplier:  Amarell
Frequently Bought Together