Order Entry
United Kingdom
ContactUsLinkComponent
Precision therm., similar ISO 655, long, -15+45:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, max. 375x9mm, richter-top finish with button, null
Catalog # AMARN65044-CB
Supplier:  Amarell
Precision therm., similar ISO 655, long, -15+45:0,2C, enclosed scale, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, max. 375x9mm, richter-top finish with button, null
Catalog # AMARN65044-CB
Supplier:  Amarell
Frequently Bought Together