Order Entry
United Kingdom
ContactUsLinkComponent
Precision therm., similar ISO 655, long, +60+85:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, max. 375x9mm, richter-top finish with button, with ice point scale, null
Catalog # AMARN65008-CB
Supplier:  Amarell
Precision therm., similar ISO 655, long, +60+85:0,1°C, enclosed scale, capillary tube specially coated inside, with non-wetting blu e special liquid, total immersion, max. 375x9mm, richter-top finish with button, with ice point scale, null
Catalog # AMARN65008-CB
Supplier:  Amarell
Frequently Bought Together