Order Entry
United Kingdom
ContactUsLinkComponent
Precision thermometer made of Quartz-Glass, -10+360:1°C, 400x8-9mm, enclosed scale, cap. colourless, gallium filled, pint-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ146008-GA
Supplier:  Amarell
Precision thermometer made of Quartz-Glass, -10+360:1°C, 400x8-9mm, enclosed scale, cap. colourless, gallium filled, pint-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARQ146008-GA
Supplier:  Amarell
Frequently Bought Together