Order Entry
United Kingdom
ContactUsLinkComponent
Extreme precision therm., with saddle, 0+100:0,1°C, enclosed scale, capillar tube specially coated inside, with non-wetting blue sp ecial liquid, 580x9-10mm, total immersion, richter-top finish with button, null
Catalog # AMARL53170-CB
Supplier:  Amarell
Extreme precision therm., with saddle, 0+100:0,1°C, enclosed scale, capillar tube specially coated inside, with non-wetting blue sp ecial liquid, 580x9-10mm, total immersion, richter-top finish with button, null
Catalog # AMARL53170-CB
Supplier:  Amarell
Frequently Bought Together