Order Entry
United Kingdom
ContactUsLinkComponent
Thermometer similar to IP 43 C, solid stem, white backed, 10+110:0,5°C, capillary tube specially coated inside, with non-wetting bl ue special liquid, durable pigment, null
  AMARA340600-CB
 :  Amarell
Thermometer similar to IP 43 C, solid stem, white backed, 10+110:0,5°C, capillary tube specially coated inside, with non-wetting bl ue special liquid, durable pigment, null
  AMARA340600-CB
 :  Amarell
Frequently Bought Together