Order Entry
United Kingdom
ContactUsLinkComponent
Thermometer similar to IP 42 C, solid stem, white backed, -38+30:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, immersion 250mm, 370x6-8mm, null
Catalog # AMARA340590-CB
Supplier:  Amarell
Thermometer similar to IP 42 C, solid stem, white backed, -38+30:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, immersion 250mm, 370x6-8mm, null
Catalog # AMARA340590-CB
Supplier:  Amarell
Frequently Bought Together