Order Entry
United Kingdom
ContactUsLinkComponent
Thermometer similar to IP 39 C, solid stem, white backed, -1+38:0,1C, capillary tube specially coated inside, with non-wetting blu e special liquid, durable pigment, null
  AMARA340550-CB
 :  Amarell
Thermometer similar to IP 39 C, solid stem, white backed, -1+38:0,1C, capillary tube specially coated inside, with non-wetting blu e special liquid, durable pigment, null
  AMARA340550-CB
 :  Amarell
Frequently Bought Together