Precision therm., straight stem, -20+360:2°C, cap. lens front, colourless, Eco-friendly, green liquid, biodegradable, upper part 275 x11mm, lower part 200x8mm, suitable for government calibration 1 * 1 items
Catalog # AMARL36776-EF
Supplier: Amarell
Frequently Bought Together